Learn More
Invitrogen™ ZNF38 Monoclonal Antibody (4B11)

Description
Peptide Sequence: MMTKVLGMAP VLGPRPPQEQ VGPLMVKVEE KEEKGKYLPS LEMFRQRFRQ FGYHDTPGPR EALSQLRVLC CEWLRPEIHT KEQILELLVL EQFLTILPQE LQAWVQEHCP ESAEEAVTLL EDLERELDEP GHQVSTPPNE QKPVWEKISS SGTAKESPSS MQPQPLETSH KYESWGPLYI QESGEEQEFA QDPRKVRDCR LSTQHEESAD EQKGSEAEGL KGDIISVIIA NKPEASLERQ CVNLENEKGT KPPLQEAGSK KGRESVPTKP TPGERRYICA ECGKAFSNSS NLTKHRRTHT GEKPYVCTKC GKAFSHSSNL TLHYRTHLVD RPYDCKCGKA FGQSSDLLKH QRMHTEEAPY QCKDCGKAFS GKGSLIRHYR IHTGEKPYQC NECGKSFSQH AGLSSHQRLH TGEKPYKCKE CGKAFNHSSN FNKHHRIHTG EKPYWCHHCG KTFCSKSNLS KHQRVHTGEG EAP.
Specifications
Specifications
| Antigen | ZNF38 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4B11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZSCAN21 |
| Gene Accession No. | Q9Y5A6 |
| Gene Alias | AI326272; CTfin51; hypothetical protein LOC527103; NY-REN-21; renal carcinoma antigen NY-REN-21; RU49; Transcription factor RU49; Zfp38; zfp-38; zinc finger and SCAN domain containing 21; zinc finger and SCAN domain-containing protein 21; zinc finger and SCAN domain-containing protein 21; zinc finger protein 3; zinc finger protein with KRAB and SCAN domains 1; zinc finger proliferation 1; zinc finger protein 38; zinc finger protein 38 (KOX 25); zinc finger protein 38 homolog; zinc finger protein NY-REN-21 antigen; zinc finger with KRAB and SCAN domains 1; Zipro1; ZKSCAN1; ZNF38; ZSCAN21 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.