Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF38 Monoclonal Antibody (OTI2E5)
GREENER_CHOICE

Catalog No. PIMA526251
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526251 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526251 Supplier Invitrogen™ Supplier No. MA526251
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF38 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

ZSCAN21 belongs to the krueppel C2H2-type zinc-finger protein family and is a strong transcriptional activator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF38
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI2E5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZSCAN21
Gene Accession No. Q07231, Q9Y5A6
Gene Alias AI326272; CTfin51; hypothetical protein LOC527103; NY-REN-21; renal carcinoma antigen NY-REN-21; RU49; Transcription factor RU49; Zfp38; zfp-38; zinc finger and SCAN domain containing 21; zinc finger and SCAN domain-containing protein 21; zinc finger and SCAN domain-containing protein 21; zinc finger protein 3; zinc finger protein with KRAB and SCAN domains 1; zinc finger proliferation 1; zinc finger protein 38; zinc finger protein 38 (KOX 25); zinc finger protein 38 homolog; zinc finger protein NY-REN-21 antigen; zinc finger with KRAB and SCAN domains 1; Zipro1; ZKSCAN1; ZNF38; ZSCAN21
Gene Symbols ZSCAN21
Host Species Mouse
Immunogen Full length human recombinant protein of ZSCAN21 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22697, 7589
Target Species Human, Mouse, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.