Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF382 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541652
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541652 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541652 Supplier Invitrogen™ Supplier No. PA541652
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MPLQGSVSFK DVTVDFTQEE WQQLDPAQKA LYRDVMLENY CHFVSVGFHM Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 92%; Rat: 93%.

Members of the C2H2 zinc finger transcription factor family, such as ZNF382, play key roles in the regulation of cell proliferation, differentiation, and apoptosis in response to a variety of stimuli. Members of the C2H2 zinc finger transcription factor family, such as ZNF382, play key roles in the regulation of cell proliferation, differentiation, and apoptosis in response to a variety of stimuli (Gebelein et al., 1998).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF382
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Znf382
Gene Accession No. Q96SR6
Gene Alias 5930415A09Rik; KRAB/zinc finger suppressor protein 1; KRAB/zinc finger suppressor protein 1 (KS1); KS1; LOW QUALITY PROTEIN: zinc finger protein 382; multiple zinc finger and krueppel-associated box protein KS1; Unknown (protein for IMAGE:8054130); Zfp382; Zinc finger protein 382; ZNF382
Gene Symbols Znf382
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF382 (aa 1-50).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84911
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.