Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF382 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561920
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561920 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561920 Supplier Invitrogen™ Supplier No. PA561920
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEQPFDHNEC EKSFLMKGML FTHTRAHRGE RTFEYNKDGI AFIEKSSLSV HPSNLMEKKP SAYNKYGKFL CRKPV Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 64%.

Members of the C2H2 zinc finger transcription factor family, such as ZNF382, play key roles in the regulation of cell proliferation, differentiation, and apoptosis in response to a variety of stimuli. Members of the C2H2 zinc finger transcription factor family, such as ZNF382, play key roles in the regulation of cell proliferation, differentiation, and apoptosis in response to a variety of stimuli (Gebelein et al., 1998).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF382
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Znf382
Gene Accession No. Q96SR6
Gene Alias 5930415A09Rik; KRAB/zinc finger suppressor protein 1; KRAB/zinc finger suppressor protein 1 (KS1); KS1; LOW QUALITY PROTEIN: zinc finger protein 382; multiple zinc finger and krueppel-associated box protein KS1; Unknown (protein for IMAGE:8054130); Zfp382; Zinc finger protein 382; ZNF382
Gene Symbols Znf382
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF382. Recombinant protein control fragment (Product #RP-100977).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84911
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.