Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF384 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552044 Supplier Invitrogen™ Supplier No. PA552044
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SHFNSNPYFW PSIPTVSGQI ENTMFINKMK DQLLPEKGCG LAPPHYPTLL TVPASVSLPS GISMDTESKS DQLTPHSQAS VTQNITVVPV PSTGLMTA Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

This gene contains long CAG trinucleotide repeats coding consecutive glutamine residues. The gene product may functions as a transcription factor, with a potential role in the regulation of neurodevelopment or neuroplasticity. The protein appears to bind and regulate the promoters of MMP1, MMP3, MMP7 and COL1A1. Studies in mouse suggest that nuclear matrix transcription factors (NP/NMP4) may be part of a general mechanical pathway that couples cell construction and function during extracellular matrix remodeling. Multiple transcript variants encoding several isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF384
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF384
Gene Accession No. Q8TF68
Gene Alias BB163993; C130073D16Rik; CAG repeat protein 1; CAGH1; CAGH1A; Cas-associated zinc finger protein; Cas-interacting zinc finger protein; Ciz; ERDA2; expanded repeat domain, CAG/CTG 2; NMP4; NP; nuclear matrix protein 4; Nuclear matrix transcription factor 4; TNRC1; Trinucleotide repeat-containing gene 1 protein; Zfp384; Zinc finger protein 384; Znf384
Gene Symbols ZNF384
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF384. Recombinant protein control fragment (Product #RP-102311).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171017
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.