Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF384 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527587
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527587 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527587 Supplier Invitrogen™ Supplier No. PA527587
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: MCF-7. Predicted reactivity: Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene contains long CAG trinucleotide repeats coding consecutive glutamine residues. The gene product may functions as a transcription factor, with a potential role in the regulation of neurodevelopment or neuroplasticity. The protein appears to bind and regulate the promoters of MMP1, MMP3, MMP7 and COL1A1. Studies in mouse suggest that nuclear matrix transcription factors (NP/NMP4) may be part of a general mechanical pathway that couples cell construction and function during extracellular matrix remodeling. Multiple transcript variants encoding several isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF384
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene ZNF384
Gene Accession No. Q8TF68
Gene Alias BB163993; C130073D16Rik; CAG repeat protein 1; CAGH1; CAGH1A; Cas-associated zinc finger protein; Cas-interacting zinc finger protein; Ciz; ERDA2; expanded repeat domain, CAG/CTG 2; NMP4; NP; nuclear matrix protein 4; Nuclear matrix transcription factor 4; TNRC1; Trinucleotide repeat-containing gene 1 protein; Zfp384; Zinc finger protein 384; Znf384
Gene Symbols ZNF384
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human ZNF384. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171017
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.