Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF385A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540434
Description
Peptide sequence: LKQHISSRRH RDGVAGKPNP LLSRHKKSRG AGELAGTLTF SKELPKSLAG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
ZNF385 is a new candidate transcription factor.
Specifications
ZNF385A | |
Polyclonal | |
Unconjugated | |
ZNF385A | |
G-protein coupled receptor 84; zinc finger protein 385A; hematopoietic zinc finger protein; Hzf; LOW QUALITY PROTEIN: zinc finger protein 385A; retinal zinc finger protein; RZF; si:dkey-228g21.6; wu:fj44b04; ZFP385; Zfp385a; zinc finger protein 385; zinc finger protein 385A; ZNF385; Znf385a | |
Rabbit | |
Affinity Chromatography | |
RUO | |
25946 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96PM9 | |
ZNF385A | |
Synthetic peptide directed towards the C-terminal of human ZNF385 (aa 276-325). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction