Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF385A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145133
Description
Immunogen sequence: PVSMENGLGP APGSPEKQPG SPSPPSIPET GQGVTKGEGG TPAPASLPGG SKEEE Antigen sequence identity: Mouse 93%; Rat 93%.
ZNF385 is a new candidate transcription factor.
Specifications
ZNF385A | |
Polyclonal | |
Unconjugated | |
ZNF385A | |
G-protein coupled receptor 84; zinc finger protein 385A; hematopoietic zinc finger protein; Hzf; LOW QUALITY PROTEIN: zinc finger protein 385A; retinal zinc finger protein; RZF; si:dkey-228g21.6; wu:fj44b04; ZFP385; Zfp385a; zinc finger protein 385; zinc finger protein 385A; ZNF385; Znf385a | |
Rabbit | |
Affinity chromatography | |
RUO | |
25946 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96PM9 | |
ZNF385A | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 140-194. Recombinant protein control fragment (Product #RP-109894). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction