Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF385B Monoclonal Antibody (OTI1B3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA802758

Description
ZNF385B contains 4 U1-type zinc fingers. ZNF385B is expressed in brain with a potential role in RNA maturation, stability, or transcription regulation. There are 5 known isoforms.Specifications
ZNF385B | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q569K4 | |
ZNF385B | |
Human recombit protein fragment corresponding to amino acids 149-399 of human ZNF385B produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI1B3 | |
Unconjugated | |
ZNF385B | |
ZNF533 | |
Mouse | |
Affinity Chromatography | |
RUO | |
151126 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction