Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF385B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565425
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565425 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565425 Supplier Invitrogen™ Supplier No. PA565425
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSKHAKKVKAL DATKNKPKMV PSKDSAKANP SCSITPITGN NSDKSEDKGK LKASSSSQP Highest antigen sequence identity to the following orthologs - mouse 88%, rat 87%.

ZNF385B contains 4 U1-type zinc fingers. ZNF385B is expressed in brain with a potential role in RNA maturation, stability, or transcription regulation. There are 5 known isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF385B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF385B
Gene Accession No. Q569K4
Gene Alias B830010L13Rik; C130013B13Rik; si:busm1-42h5.1; si:by51i9.1; si:dz145b16.1; si:dz42h5.1; si:xx-51i9.1; wu:fj47d04; ZFN533; Zfp385b; Zfp533; zgc:73248; Zinc finger protein 385B; zinc finger protein 533; ZNF385B; ZNF533
Gene Symbols ZNF385B
Host Species Rabbit
Immunogen Recombinant Human ZNF385B. Recombinant protein control fragment (Product #RP-104813).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151126
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.