Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF385C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571541
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571541 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571541 Supplier Invitrogen™ Supplier No. PA571541
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF385C Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF385C
Applications Western Blot
Classification Polyclonal
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Zfp385c
Gene Accession No. A2A5E6, Q66K41
Gene Alias A930006D11Rik; CTD-2132N18.2; CTD-2132N18.4; Zfp385c; Zinc finger protein 385C; Znf385c
Gene Symbols Zfp385c
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 77-106 amino acids from the N-terminal region of human ZNF385C.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201181, 278304
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.