Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF385D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557566
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557566 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557566 Supplier Invitrogen™ Supplier No. PA557566
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HAKKLKALEA MKNKQKSVTA KDSAKTTFTS ITTNTINTSS DKTDGTAGTP AISTTTTVEI RKSSVMTTEI TSKVE Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 73%.

ZNF385D gene ontology annotations related to this gene include nucleic acid binding; zinc ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF385D
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF385D
Gene Accession No. Q9H6B1
Gene Alias si:dkey-82o10.2; Zfp385d; Zinc finger protein 385D; zinc finger protein 659; ZNF385D; ZNF659
Gene Symbols ZNF385D
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF385D. Recombinant protein control fragment (Product #RP-96376).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79750
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.