Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF385D Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569712
Description
This target displays homology in the following species: Dog: 93%; Horse: 93%; Human: 100%; Pig: 93%; Rabbit: 93%; Rat: 93%.
ZNF385D gene ontology annotations related to this gene include nucleic acid binding; zinc ion binding.
Specifications
ZNF385D | |
Polyclonal | |
Unconjugated | |
ZNF385D | |
si:dkey-82o10.2; Zfp385d; Zinc finger protein 385D; zinc finger protein 659; ZNF385D; ZNF659 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79750 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H6B1 | |
ZNF385D | |
synthetic peptide directed towards the N terminal region of human ZNF385D. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction