Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF391 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA562260

Catalog No. PIPA562260


Only null left
Add to Cart

Description

Description

Immunogen sequence: NTAQGPTNEE DYKNEGQLSR QTKCPAQKKS SFENTVVRKV SVTLKEIFTG EEGPESSEFS LSPNLDAQQK IPKG Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 28%.

May be involved in transcriptional regulation. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF391
Polyclonal
Unconjugated
ZNF391
dJ153G14.3; Zinc finger protein 391; ZNF391
Rabbit
Antigen Affinity Chromatography
RUO
346157
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
Lot-Specific
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9UJN7
ZNF391
Recombinant protein corresponding to Human ZNF391. Recombinant protein control fragment (Product #RP-101258).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.