Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF394 Antibody (OTI1G9), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274949AF750
Description
ZNF394 Monoclonal specifically detects ZNF394 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ZNF394 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
ZNF394 | |
Recombinant protein fragment corresponding to amino acids 1-296 of human ZNF394 (NP_115540) produced in E.coli. | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1G9 | |
Western Blot, Immunohistochemistry | |
dJ874C20.2, FLJ23407, zinc finger protein 310 pseudogene, zinc finger protein 323, ZNF20-Lp, ZNF310P | |
Mouse | |
0.1 mL | |
84124 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction