Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF394 Monoclonal Antibody (1F8)
GREENER_CHOICE

Catalog No. PIMA522729
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522729 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522729 Supplier Invitrogen™ Supplier No. MA522729
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MNSSLTAQRR GSDAELGPWV MAARSKDAAP SQRDGLLPVK VEEDSPGSWE PNYPAASPDP ETSRLHFRQL RYQEVAGPEE ALSRLRELCR RWLRPELLSK EQILELLVLE QFLTILPEEL QAWVREHCPE SGEEAVAVVR ALQRALDGTS SQGMVTFEDT AVSLTWEEWE RLDPARRDFC RESAQKDSGS TVPPSLESRV ENKELIPMQQ ILEEAEPQGQ LQEAFQGKRP LFSKCGSTHE DRVEKQSGDP LPLKLENSPE AEGLNSISDV NKNGSIEGED SKNNELQNSA RCSNLVLCQH IPKAERPTDS EEHGNKCKQS FHMVTWHVLK PHKSDSGDSF HHSSLFETQR QLHEERPYKC GNCGKSFKQR SDLFRHQRIH TGEKPYGCQE CGKSFSQSAA LTKHQRTHTG EKPYTCLKCG ERFRQNSHLN RHQSTHSRDK HFKCEECGET CHISNLFRHQ RLHKGERPYK CEECEKSFKQ RSDLFKHHRI HTGEKPYGCS VCGKRFNQSA TLIKHQRIHT GEKPYKCLEC GERFRQSTHL IRHQRIHQNK VLSAGRGGSR L.

ZNF394 belongs to the krueppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF394
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1F8
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZNF394
Gene Accession No. Q53GI3
Gene Alias 2310046C23Rik; 2810437E14Rik; Rlzfy; RLZF-Y; SNAP19; truncated zinc finger protein isoform; Unknown (protein for MGC:127479); Zfp394; Zfp94; zfp-94; Zfp99; Zinc finger protein 394; zinc finger protein 94; zinc finger protein 99; zinc finger protein LOC522836; zinc finger protein with KRAB and SCAN domains 14; Zinc finger protein Y1; zinc finger protein Y1 (RLZF-Y); zinc finger with KRAB and SCAN domains 14; ZKSCAN14; Znf394; ZSCAN46
Gene Symbols ZNF394
Host Species Mouse
Immunogen ZNF394 (AAH25241, 1 a.a. ~ 561 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84124
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.