Learn More
Invitrogen™ ZNF394 Monoclonal Antibody (OTI1G9)

Description
ZNF394 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | ZNF394 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1G9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ZNF394 |
| Gene Accession No. | Q53GI3 |
| Gene Alias | 2310046C23Rik; 2810437E14Rik; Rlzfy; RLZF-Y; SNAP19; truncated zinc finger protein isoform; Unknown (protein for MGC:127479); Zfp394; Zfp94; zfp-94; Zfp99; Zinc finger protein 394; zinc finger protein 94; zinc finger protein 99; zinc finger protein LOC522836; zinc finger protein with KRAB and SCAN domains 14; Zinc finger protein Y1; zinc finger protein Y1 (RLZF-Y); zinc finger with KRAB and SCAN domains 14; ZKSCAN14; Znf394; ZSCAN46 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.