Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF394 Monoclonal Antibody (OTI1H10)
GREENER_CHOICE

Catalog No. PIMA527075
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527075 Supplier Invitrogen™ Supplier No. MA527075
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF394 Monoclonal Antibody for Western Blot, IHC (P)

ZNF394 belongs to the krueppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF394
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1H10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZNF394
Gene Accession No. Q53GI3
Gene Alias 2310046C23Rik; 2810437E14Rik; Rlzfy; RLZF-Y; SNAP19; truncated zinc finger protein isoform; Unknown (protein for MGC:127479); Zfp394; Zfp94; zfp-94; Zfp99; Zinc finger protein 394; zinc finger protein 94; zinc finger protein 99; zinc finger protein LOC522836; zinc finger protein with KRAB and SCAN domains 14; Zinc finger protein Y1; zinc finger protein Y1 (RLZF-Y); zinc finger with KRAB and SCAN domains 14; ZKSCAN14; Znf394; ZSCAN46
Gene Symbols ZNF394
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-296 of ZNF394 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84124
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.