Learn More
ZNF394 Mouse anti-Human, Clone: OTI3E4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808958

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ZNF394 belongs to the krueppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation.Specifications
ZNF394 | |
Monoclonal | |
Unconjugated | |
ZNF394 | |
ZKSCAN14, ZSCAN46 | |
Mouse | |
Affinity Chromatography | |
RUO | |
84124 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3E4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q53GI3 | |
ZNF394 | |
Human recombit protein fragment corresponding to amino acids 1-296 of human ZNF394 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.