Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ ZNF394 Polyclonal Antibody
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562996
Description
Immunogen sequence: QGMVTFEDTA VSLTWEEWER LDPARRDFCR ESAQKDSGST VPPSLESRVE NKELIPMQQI LEEAEPQGQL QEAFQGKRPL FSKCGSTH Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 55%.
ZNF394 belongs to the krueppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation. 
            Specifications
| ZNF394 | |
| Polyclonal | |
| Unconjugated | |
| ZNF394 | |
| 2310046C23Rik; 2810437E14Rik; Rlzfy; RLZF-Y; SNAP19; truncated zinc finger protein isoform; Unknown (protein for MGC:127479); Zfp394; Zfp94; zfp-94; Zfp99; Zinc finger protein 394; zinc finger protein 94; zinc finger protein 99; zinc finger protein LOC522836; zinc finger protein with KRAB and SCAN domains 14; Zinc finger protein Y1; zinc finger protein Y1 (RLZF-Y); zinc finger with KRAB and SCAN domains 14; ZKSCAN14; Znf394; ZSCAN46 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84124 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | 
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q53GI3 | |
| ZNF394 | |
| Recombinant protein corresponding to Human ZNF394. Recombinant protein control fragment (Product #RP-98509). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                             
                            