Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF394 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569033
Description
This target displays homology in the following species: Cow: 92%; Dog: 92%; Guinea Pig: 83%; Horse: 92%; Human: 100%; Mouse: 93%; Pig: 92%; Rabbit: 100%; Rat: 100%.
ZNF394 belongs to the krueppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation.
Specifications
ZNF394 | |
Polyclonal | |
Unconjugated | |
Zkscan14 | |
2310046C23Rik; 2810437E14Rik; Rlzfy; RLZF-Y; SNAP19; truncated zinc finger protein isoform; Unknown (protein for MGC:127479); Zfp394; Zfp94; zfp-94; Zfp99; Zinc finger protein 394; zinc finger protein 94; zinc finger protein 99; zinc finger protein LOC522836; zinc finger protein with KRAB and SCAN domains 14; Zinc finger protein Y1; zinc finger protein Y1 (RLZF-Y); zinc finger with KRAB and SCAN domains 14; ZKSCAN14; Znf394; ZSCAN46 | |
Rabbit | |
Affinity chromatography | |
RUO | |
67235 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Z1D9 | |
Zkscan14 | |
synthetic peptide directed towards the middle region of human ZNF394 (aa 471-520). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction