Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF395 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 117591AP150UL

Description
Papillomavirus Regulatory Factor 1/HDBP2 is a novel transcription factor shuttling between nucleus and cytoplasm and binds to the specific GCCGGCG, which is an essential cis-element for Huntington′s disease gene expression.Specifications
ZNF395 | |
Polyclonal | |
Unconjugated | |
ZNF395 | |
HD regulating factor 2, HDBP 2, HDBP2, HDRF 2, Papillomavirus binding factor, PBF, PRF 1, PRF1, Si 1 8 14, zinc finger protein 395, ZNF395 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55893 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H8N7 | |
ZNF395 | |
ZNF395 Fusion Protein Ag2355 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction