Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF396 Monoclonal Antibody (2F8)
GREENER_CHOICE

Catalog No. PIMA523205
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523205 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523205 Supplier Invitrogen™ Supplier No. MA523205
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: ERELDGPKQI FFGRRKDMIA EKLAPSEITE ELPSSQLMPV KKQLQGASWE LQSLRPHDED IKTTNVKSAS RQKTSLGIEL HCNVSNILHM NGSQSSTYRG.

Isoform 1 and isoform 2 act as DNA-dependent transcriptional repressors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF396
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2F8
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZNF396
Gene Accession No. Q96N95
Gene Alias Zinc finger and SCAN domain-containing protein 14; zinc finger protein 396; ZNF396; ZSCAN14
Gene Symbols ZNF396
Host Species Mouse
Immunogen ZNF396 (NP_665699.1, 131 a.a. ~ 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 252884
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.