Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF396 Polyclonal Antibody
ZNF396 Polyclonal Antibody
Supplier: Thermo Scientific PA582258
Description
ZNF396 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
ZNF396 | |
Polyclonal | |
Unconjugated | |
ZNF396 | |
Zinc finger and SCAN domain-containing protein 14; zinc finger protein 396; ZNF396; ZSCAN14 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
252884 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96N95 | |
ZNF396 | |
Recombinant protein corresponding to Human ZNF396. Recombinant protein control fragment (Product #RP-89422). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction