Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF397 Antibody (OTI1F3), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274950C
Description
ZNF397 Monoclonal specifically detects ZNF397 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ZNF397 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ZNF397 | |
Full length recombinant protein of human ZNF397 (NP_115723) produced in HEK293T cells. | |
0.1 mL | |
84307.0 | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1F3 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
KIAA1588, KRAB-containing zinc finger protein 317, zinc finger protein 317 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction