Learn More
Invitrogen™ ZNF397 Monoclonal Antibody (1F8)

Description
Peptide Sequence: EFDDPGQQVP ASPQGPAVPW KDLTCLRASQ ESTDIHLQPL KTQLKSWKPC LSPKSDCENS ETATKEGISE EKSQGLPQEP SFRGIKLSRP PKASSAIRWE.
Specifications
Specifications
| Antigen | ZNF397 |
| Applications | ELISA, Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1F8 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZNF397 |
| Gene Accession No. | Q8NF99 |
| Gene Alias | 2810411K16Rik; 6720480F11Rik; AI461664; Zfp397; Zinc finger and SCAN domain-containing protein 15; zinc finger protein 397; Zinc finger protein 47; ZNF397; ZNF47; ZSCAN15 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.