Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF397 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541647
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541647 Supplier Invitrogen™ Supplier No. PA541647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SAIRWECVSP GSFPGDIIAA EATHSTISCF AINTLPATIL PSKNVNRKYF Sequence homology: Human: 100%.

This gene encodes a protein with a N-terminal SCAN domain, and the longer isoform contains nine C2H2-type zinc finger repeats in the C-terminal domain. The protein localizes to centromeres during interphase and early prophase, and different isoforms can repress or activate transcription in transfection studies. Multiple transcript variants encoding different isoforms have been found for this gene. Additional variants have been described, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF397
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF397
Gene Accession No. Q8NF99
Gene Alias 2810411K16Rik; 6720480F11Rik; AI461664; Zfp397; Zinc finger and SCAN domain-containing protein 15; zinc finger protein 397; Zinc finger protein 47; ZNF397; ZNF47; ZSCAN15
Gene Symbols ZNF397
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ZNF397.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84307
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.