Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF397 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582902
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582902 Supplier Invitrogen™ Supplier No. PA582902
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QQHNPESGEE AVTLLEDLER EFDDPGQQVP ASPQGPAVPW KDLTCLRASQ ESTDIHLQPL KTQLKSWKPC LSPKSDCENS ETATKEGISE EKSQGLPQEP SFRGIKLSRP PKASSAIRWE CVSPGSFPGD IIAAEATHST ISCFAINTL Highest antigen sequence indentity to the following orthologs: Mouse - 52%, Rat - 54%.

This gene encodes a protein with a N-terminal SCAN domain, and the longer isoform contains nine C2H2-type zinc finger repeats in the C-terminal domain. The protein localizes to centromeres during interphase and early prophase, and different isoforms can repress or activate transcription in transfection studies. Multiple transcript variants encoding different isoforms have been found for this gene. Additional variants have been described, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF397
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF397
Gene Accession No. Q8NF99
Gene Alias 2810411K16Rik; 6720480F11Rik; AI461664; Zfp397; Zinc finger and SCAN domain-containing protein 15; zinc finger protein 397; Zinc finger protein 47; ZNF397; ZNF47; ZSCAN15
Gene Symbols ZNF397
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF397. Recombinant protein control fragment (Product #RP-89423).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84307
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.