Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF398 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145042
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145042 Supplier Invitrogen™ Supplier No. PA5145042
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QLGSYPLPPP VGEQVFSCHH CGKNLSQDML LTHQCSHATE HPLPCAQCPK HFTPQADLSS TSQDHASE Antigen sequence identity: Mouse 88%; Rat 93%.

Function as a transcriptional activator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF398
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF398
Gene Accession No. Q8TD17
Gene Alias KIAA1339; P51; P71; ZER6; zinc finger DNA binding protein p52/p71; zinc finger DNA binding protein ZER6; zinc finger DNA-binding protein p52/p71; Zinc finger protein 398; zinc finger-estrogen receptor interaction, clone 6; ZNF398
Gene Symbols ZNF398
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 328-395. Recombinant protein control fragment (Product #RP-109851).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57541
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.