Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    ZNF407 Antibody, HRP, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306058H
Description
ZNF407 Polyclonal antibody specifically detects ZNF407 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| ZNF407 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| FLJ13839, FLJ20307, KIAA1703, zinc finger protein 407 | |
| Produced in rabbits immunized with E. coli-derived Human ZNF407 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified | 
| Immunocytochemistry | |
| HRP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 55628 | |
| Store at 4C in the dark. | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            