Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF408 Antibody (PCRP-ZNF408-1E5), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308324MFV500
Description
ZNF408 Monoclonal antibody specifically detects ZNF408 in Human samples. It is validated for Flow CytometrySpecifications
ZNF408 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
79797 | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry | |
PCRP-ZNF408-1E5 | |
Flow Cytometry | |
A8-51, C2-H2 type zinc finger protein, FLJ20216, HF.12, HZF3.1, KOX25zinc finger protein 3 (A8-51), PP838, Zfp113, zinc finger protein 3, Zinc finger protein HF.12, Zinc finger protein HZF3.1, Zinc finger protein KOX25 | |
Recombinant full-length human ZNF408 protein (Uniprot: Q9H9D4) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction