Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF408 Mouse anti-Human, Clone: PCRP-ZNF408-1E5, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | ZNF408 |
---|---|
Clone | PCRP-ZNF408-1E5 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Protein Array |
Applications | Western Blot, Flow Cytometry, Peptide Array |
Description
ZNF408 Monoclonal specifically detects ZNF408 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein Array.Specifications
ZNF408 | |
0.2 mg/ml | |
Western Blot, Flow Cytometry, Peptide Array | |
Unconjugated | |
Mouse | |
Human | |
A8-51, C2-H2 type zinc finger protein, FLJ20216, HF.12, HZF3.1, KOX25zinc finger protein 3 (A8-51), PP838, Zfp113, zinc finger protein 3, Zinc finger protein HF.12, Zinc finger protein HZF3.1, Zinc finger protein KOX25 | |
Recombinant full-length human ZNF408 protein | |
Primary | |
Store at 4°C. |
PCRP-ZNF408-1E5 | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
79797 | |
IgG2b | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title