Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF408 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553660
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553660 Supplier Invitrogen™ Supplier No. PA553660
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AAVAVVTEVE SAVQQEVASP GEDAAEPCID PGSQSPSGIQ AENMVSPGLK FPTQDRISKD SQPLGPLLQD GDVDEECPAQ AQMPPELQSN SATQQDPDGS GASFSSSARG TQPHGYLAKK LHSPSDQCPP RAKTPEPGAQ QSGFPTLSRS Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 25%.

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. As a member of the Kruppel C2H2-type zinc-finger protein family, ZNF396 (zinc finger protein 396), also known as PRDM17 (PR domain zinc finger protein 17), is a 720 amino acid nuclear protein that contains 10 C2H2-type zinc fingers. The gene encoding ZNF408 maps to human chromosome 11, which houses over 1,400 genes and comprises nearly 4% of the human genome. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are associated with defects in genes that maps to chromosome 11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF408
Applications ChIP Assay, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF408
Gene Accession No. Q9H9D4
Gene Alias EVR6; LOC100362202; PFM14; PR domain zinc finger protein 17; PRDM17; RP72; Zinc finger protein 408; ZNF408
Gene Symbols ZNF408
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF408. Recombinant protein control fragment (Product #RP-91666).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79797
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.