Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF408 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576410
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. As a member of the Kruppel C2H2-type zinc-finger protein family, ZNF396 (zinc finger protein 396), also known as PRDM17 (PR domain zinc finger protein 17), is a 720 amino acid nuclear protein that contains 10 C2H2-type zinc fingers. The gene encoding ZNF408 maps to human chromosome 11, which houses over 1,400 genes and comprises nearly 4% of the human genome. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are associated with defects in genes that maps to chromosome 11.
Specifications
ZNF408 | |
Polyclonal | |
Unconjugated | |
ZNF408 | |
EVR6; LOC100362202; PFM14; PR domain zinc finger protein 17; PRDM17; RP72; Zinc finger protein 408; ZNF408 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100362202, 79797 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H9D4 | |
ZNF408 | |
Recombinant fusion protein of human ZNF408. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction