Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF410 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP221008
Description
ZNF410 Polyclonal specifically detects ZNF410 in Human samples. It is validated for Western Blot.Specifications
ZNF410 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
DKFZp781N1974, gene similar to zinc finger 5 protein10, ZBTB21KIAA1227, Zinc finger and BTB domain-containing protein 21, zinc finger protein 295 | |
Rabbit | |
52 kDa | |
0.1 mL | |
Primary | |
Human. Species reactivity based on sequence identity: Chimpanzee (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
Q86VK4 | |
ZNF410 | |
Recombinant protein encompassing a sequence within the center region of human ZNF410. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
57862 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction