Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF410 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531252
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Chicken (84%), Chimpanzee (100%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Transcription factor that activates transcription of matrix-remodeling genes such as MMP1 during fibroblast senescence.
Specifications
ZNF410 | |
Polyclonal | |
Unconjugated | |
ZNF410 | |
Another partner for ARF 1; Apa1; APA-1; clones 23667 and 23775 zinc finger protein; D12Ertd748e; Zfp410; Zinc finger protein 410; zinc finger protein APA-1; Znf410 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57862 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q86VK4 | |
ZNF410 | |
Recombinant fragment corresponding to a region within amino acids 36 and 261 of Human ZNF410. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction