Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF410 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501729519 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9519 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9519 Supplier Proteintech Group Inc Supplier No. 145291AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transcription factor that activates transcription of matrix-remodeling genes such as MMP1 during fibroblast senescence.

Specifications

Antigen ZNF410
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZNF410
Gene Accession No. Q86VK4, Q8BKX7
Gene Alias Another partner for ARF 1, APA 1, APA1, zinc finger protein 410, Zinc finger protein APA 1, ZNF410
Gene Symbols Zfp410, Znf410
Host Species Rabbit
Immunogen ZNF410 Fusion Protein Ag6028
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 314310, 52708, 57862
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.