Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF415 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ZNF415 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ZNF415 Polyclonal specifically detects ZNF415 in Human samples. It is validated for Western Blot.Specifications
ZNF415 | |
Polyclonal | |
Rabbit | |
NP_060825 | |
55786 | |
Synthetic peptide directed towards the C terminal of human ZNF415. Peptide sequence SVRPNLTRHQIIHTGKKPYKCSDCGKSFSVRPNLFRHQIIHTKEKPYKRN. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
MGC126536, MGC141923, zinc finger protein 287, ZKSCAN13Zinc finger protein with KRAB and SCAN domains 13 | |
ZNF415 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title