Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF415 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987944
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7944 Supplier Bioss Supplier No. BS7708R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF415 is involved in transcriptional regulation. The transcriptional activity differed among the various isoforms. All isoforms except isoform 3 seem to suppress the transcriptional activities of AP-1 and p53.

Specifications

Antigen ZNF415
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZNF415
Gene Accession No. Q09FC8
Gene Alias ZfLp; Zinc finger protein 415; ZNF415
Gene Symbols ZNF415
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZNF415.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55786
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.