Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF416 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589809
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589809 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589809 Supplier Invitrogen™ Supplier No. PA589809
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ECSECGKFFR QSYTLVEHQK IHTGLRPYDC GQCGKSFIQK SSLIQHQVVH TGERPYECGK CGKSFTQHSG LILHRKSHTV ERPRDSSKCG KPYSPRSNIV; Positive Samples: Mouse kidney; Cellular Location: Nucleus.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF416
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Alias Zinc finger protein 416; ZNF416
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 495-594 of human ZNF416 (NP_0603491).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.