Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF814 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562835
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562835 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562835 Supplier Invitrogen™ Supplier No. PA562835
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGTHHKQKLH RCEAWGNKLY DSGNFHQHQ Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 45%.

Predicted to enable DNA-binding transcription activator activity, RNA polymerase II-specific and RNA polymerase II cis-regulatory region sequence-specific DNA binding activity. Predicted to be involved in regulation of transcription by RNA polymerase II. Predicted to be active in nucleus. [provided by Alliance of Genome Resources, Apr 2022].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF814
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF814
Gene Accession No. B7Z6K7
Gene Alias FLJ31551; KIAA1956; MGC138449; Zinc finger protein 418; ZNF418
Gene Symbols ZNF814
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF418.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 730051
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.