Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF423 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB423771 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB423771 0.02 mL
NB423770 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB423771 Supplier Novus Biologicals Supplier No. NBP2941620.02ML

Rabbit Polyclonal Antibody

ZNF423 Polyclonal antibody specifically detects ZNF423 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen ZNF423
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias FLJ20086, KIAA1584, MGC21637, MGC61687, SUHW4, Suppressor of hairy wing homolog 4, zinc finger protein 280D, Zinc finger protein 634, ZNF634suppressor of hairy wing homolog 4 (Drosophila)
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 220-450 of human ZNF423 (NP_055884.2). KPFKCTVCKRGFSSTSSLQSHMQAHKKNKEHLAKSEKEAKKDDFMCDYCEDTFSQTEELEKHVLTRHPQLSEKADLQCIHCPEVFVDENTLLAHIHQAHANQKHKCPMCPEQFSSVEGVYCHLDSHRQPDSSNHSVSPDPVLGSVASMSSATPDSSASVERGSTPDSTLKPLRGQKKMRDDGQGWTKVVYSCPYCSKRDFNSLAVLEIHLKTIHADKPQQSHTCQICLDSM
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23090
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.