Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF423/OAZ Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982047
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2047 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2047 Supplier Bioss Supplier No. BS12220R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transcription factor that can both act as an activator or a repressor depending on the context. Plays a central role in BMP signaling and olfactory neurogenesis. Associates with SMADs in response to BMP2 leading to activate transcription of BMP target genes. Acts as a transcriptional repressor via its interaction with EBF1, a transcription factor involved in terminal olfactory receptor neurons differentiation; this interaction preventing EBF1 to bind DNA and activate olfactory-specific genes. Involved in olfactory neurogenesis by participating in a developmental switch that regulates the transition from differentiation to maturation in olfactory receptor neurons. Controls proliferation and differentiation of neural precursors in cerebellar vermis formation.

Specifications

Antigen ZNF423/OAZ
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZNF423
Gene Accession No. Q2M1K9
Gene Alias early B-cell factor associated zinc finger protein; Ebfaz; hOAZ; JBTS19; KIAA0760; NPHP14; OAZ; OLF-1/EBF associated zinc finger; Olf1/EBF-associated zinc finger protein; Roaz; Smad- and Olf-interacting zinc finger protein; Zfp104; ZFP423; Zinc finger protein 423; ZNF423
Gene Symbols ZNF423
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZNF423/OAZ.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23090
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.