Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF426 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556117
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556117 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556117 Supplier Invitrogen™ Supplier No. PA556117
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EKTPAGRIVA DCLTDCYQDS VTFDDVAVDF TQEEWTLLDS TQRSLYSDVM LENYKNLATV GGQIIKPSLI SWLEQEESRT VQGGVLQGWE MRLETQWSIL QQDFLRGQTS IGIQLEGKHN GRELC Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 62%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF426
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF426
Gene Accession No. Q9BUY5
Gene Alias CTC-543D15.7; K-RBP; KSHV RTA binding protein; zinc finger protein 426; ZNF426
Gene Symbols ZNF426
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF426. Recombinant protein control fragment (Product #RP-99690).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79088
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.