Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF426 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110601
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110601 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110601 Supplier Invitrogen™ Supplier No. PA5110601
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QCGEVFSEHS CLKTHVRTQS TGNTHDCNQY GKDFLTLCEK TSTGEKLSEF NQSEKIFSLT PNIVYQRTST QEKSFECSHC GKSFINESYL QAHMRTHNGE KLYEWRNYGP GFIDSTSLSV L.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF426
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene ZNF426
Gene Accession No. Q9BUY5
Gene Alias CTC-543D15.7; K-RBP; KSHV RTA binding protein; zinc finger protein 426; ZNF426
Gene Symbols ZNF426
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 150-270 of human ZNF426 (NP_077011.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79088
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.