Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF428 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571584
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571584 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571584 Supplier Invitrogen™ Supplier No. PA571584
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF428 Polyclonal Antibody for Western Blot

ZNF428 contains 1 C2H2-type zinc finger. The exact function of ZNF428 remains unknown. Two different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF428
Applications Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ZNF428
Gene Accession No. Q8C1M2, Q96B54
Gene Alias 2410005H09Rik; AV271806; AW047854; C19orf37; Enzyme-like protein PIT13; Zfp428; Zinc finger protein 428; Znf428
Gene Symbols Zfp428, ZNF428
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 42-70 amino acids from the N-terminal region of human ZNF428.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126299, 232969
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.