Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF428 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541695
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541695 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541695 Supplier Invitrogen™ Supplier No. PA541695
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PCRLCCPATA PQEAPAPEGR ALGEEEEEPP RAGEGRPAGR EEEEEEEEEG Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Human: 100%; Pig: 93%; Rabbit: 100%; Rat: 88%.

ZNF428 contains 1 C2H2-type zinc finger. The exact function of ZNF428 remains unknown. Two different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF428
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF428
Gene Accession No. Q96B54
Gene Alias 2410005H09Rik; AV271806; AW047854; C19orf37; Enzyme-like protein PIT13; Zfp428; Zinc finger protein 428; Znf428
Gene Symbols ZNF428
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZNF428.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126299
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.