Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF431 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541659 Supplier Invitrogen™ Supplier No. PA541659
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TLTKHRKIHT RQKPYNCEEC DNTFNQSSNL IKQNNSYWRE TLQMSRMWES Sequence homology: Human: 100%.

ZNF431 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF431
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF431
Gene Accession No. Q8TF32
Gene Alias KIAA1969; Zinc finger protein 431; ZNF431
Gene Symbols ZNF431
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZNF431.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170959
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.