Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF432 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563830
Description
Immunogen sequence: SCEINNSTKF SGDGKSFLHG NYEELYSAAK FSVSTKANST KSQVSKHQRT HEI Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 36%.
ZNF432 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 16 C2H2-type zinc fingers and 1 KRAB domain. ZNF432 may be involved in transcriptional regulation.
Specifications
ZNF432 | |
Polyclonal | |
Unconjugated | |
ZNF432 | |
KIAA0798; Zinc finger protein 432; ZNF432 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9668 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94892 | |
ZNF432 | |
Recombinant protein corresponding to Human ZNF432. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction