Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF432 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111602
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111602 100 μL
PIPA5111346 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5111602 Supplier Invitrogen™ Supplier No. PA5111602
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SCEINNSTKF SGDGKSFLHG NYEELYSAAK FSVSTKANST KSQVSKHQRT HEI Highest antigen sequence indentity to the following orthologs: Mouse - 32%, Rat - 36%.

ZNF432 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 16 C2H2-type zinc fingers and 1 KRAB domain. ZNF432 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF432
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF432
Gene Accession No. O94892
Gene Alias KIAA0798; Zinc finger protein 432; ZNF432
Gene Symbols ZNF432
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ZNF432. Recombinant protein control fragment (Product #RP-107441).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9668
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.